6-Aminonicotinic acid 98%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
6-Aminopyridine-3-carboxylic acid, 6-Amino-3-pyridinecarboxylic acid
OR5070-100G
APOSOR5070-100GEA
1660
CZK
APOSOR5070-100G
6-Aminonicotinic acid 98%
6-Aminonicotinic acid
Vzorec:
C₆H₆N₂O₂ MW: 138,13 g/mol Bod tání: 320 °C (rozkl.) Storage Temperature: Ambient |
MDL Number:
MFCD00006326 CAS číslo: 3167-49-5 EINECS: 221-630-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...