2-Thienylacetic acid 97%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
OR5059-100G
OR5059-25G
APOSOR5059-100GEA
1700
CZK
APOSOR5059-100G
APOSOR5059-25G
2-Thienylacetic acid 97%
2-Thienylacetic acid
Vzorec:
C₆H₆O₂S Bod varu: 160 °C/22mmHg Bod tání: 61…65 °C Storage Temperature: Ambient |
MDL Number:
MFCD00005458 CAS číslo: 1918-77-0 EINECS: 217-639-8 UN: 3261 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...