2-Thenoic acid 97%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
Thiophene-2-carboxylic acid, 2-Thiophenecarboxylic acid
OR5057-100G
OR5057-25G
OR5057-500G
OR5057-1KG
APOSOR5057-100GEA
622
CZK
APOSOR5057-100G
APOSOR5057-25G
APOSOR5057-500G
APOSOR5057-1KG
2-Thenoic acid 97%
2-Thenoic acid
Vzorec:
C₅H₄O₂S MW: 128,15 g/mol Bod varu: 259…261 °C Bod tání: 126…129 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00005437 CAS číslo: 527-72-0 EINECS: 208-423-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...