Tisk…

α-Bromo-p-toluonitrile 98%

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: 4-(Bromomethyl)benzonitrile, 4-Cyanobenzyl bromide, p-(Bromomethyl)benzonitrile

OR5037-25G OR5037-100G OR5037-5G
APOSOR5037-25GEA 705 CZK
APOSOR5037-25G APOSOR5037-100G APOSOR5037-5G
α-Bromo-p-toluonitrile 98%
α-Bromo-p-toluonitrile
Vzorec: C₈H₆BrN
MW: 196.05
Bod tání: 113…117 °C
Storage Temperature: Ambient
MDL Number: MFCD00001829
CAS číslo: 17201-43-3
EINECS: 241-246-0
UN: 3261
ADR: 8 II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR