α-Bromo-2-chlorotoulene 97%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
Synonyms:
1-(Bromomethyl)-2-chlorobenzene, 2-Chlorobenzyl bromide
OR5033-100G
OR5033-25G
OR5033-5G
APOSOR5033-100GEA
3230
CZK
APOSOR5033-100G
APOSOR5033-25G
APOSOR5033-5G
α-Bromo-2-chlorotoulene 97%
α-Bromo-2-chlorotoulene
Vzorec:
C₇H₆BrCl MW: 205,48 g/mol Bod varu: 103…104 °C/10mmHg Hustota: 1,583 g/cm³ (25 °C) Bod vzplanutí: 109 °C Storage Temperature: Ambient |
MDL Number:
MFCD00000566 CAS číslo: 611-17-6 EINECS: 210-257-2 UN: 3265 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...