4-Methoxybenzylamine 97%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
Synonyms:
4-(Aminomethyl)anisole, p-Anisylamine
OR5028-100G
OR5028-25G
OR5028-500G
APOSOR5028-100GEA
1870
CZK
APOSOR5028-100G
APOSOR5028-25G
APOSOR5028-500G
4-Methoxybenzylamine 97%
4-Methoxybenzylamine
Vzorec:
C₈H₁₁NO MW: 137,18 g/mol Bod varu: 64…66 °C Hustota: 1,053 g/cm³ (25 °C) Bod vzplanutí: >110 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008122 CAS číslo: 2393-23-9 EINECS: 219-247-2 UN: 2735 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...