2-Nitrobenzyl alcohol 98%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
(2-Nitrophenyl)methanol
OR5014-100G
OR5014-500G
APOSOR5014-100GEA
1240
CZK
APOSOR5014-100G
APOSOR5014-500G
2-Nitrobenzyl alcohol 98%
2-Nitrobenzyl alcohol
Vzorec:
C₇H₇NO₃ MW: 153,14 g/mol Bod varu: 270 °C Bod tání: 69…74 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007186 CAS číslo: 612-25-9 EINECS: 210-302-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...