2,6-Dichlorobenzyl alcohol 97%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
(2,6-Dichlorophenyl)methanol
OR5013-100G
OR5013-10G
OR5013-25G
APOSOR5013-100GEA
2420
CZK
APOSOR5013-100G
APOSOR5013-10G
APOSOR5013-25G
2,6-Dichlorobenzyl alcohol 97%
2,6-Dichlorobenzyl alcohol
Vzorec:
C₇H₆Cl₂O Bod varu: 150 °C/25mmHg Bod tání: 96…99 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004609 CAS číslo: 15258-73-8 EINECS: 239-300-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...