4,4'-Dimethoxybenzophenone 98%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
Bis(4-methoxyphenyl)methanone
OR5012-100G
OR5012-25G
APOSOR5012-100GEA
1870
CZK
APOSOR5012-100G
APOSOR5012-25G
4,4'-Dimethoxybenzophenone 98%
4,4'-Dimethoxybenzophenone
Vzorec:
C₁₅H₁₄O₃ MW: 242,27 g/mol Bod tání: 141…146 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008404 CAS číslo: 90-96-0 EINECS: 202-028-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...