Tisk…

2-Bromo-6-chloroaniline 95+%

Dodavatel: Apollo Scientific
Varování

OR500007-1G OR500007-250MG OR500007-5G
APOSOR500007-1GEA 705 CZK
APOSOR500007-1G APOSOR500007-250MG APOSOR500007-5G
2-Bromo-6-chloroaniline 95+%
2-Bromo-6-chloroaniline
Vzorec: C₆H₅BrClN
MW: 206,47 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11977383
CAS číslo: 59772-49-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR