Tisk…

4-(tert-Butyl)-2-nitroaniline

Dodavatel: Apollo Scientific
Nebezpečí

OR4982-1G OR4982-5G OR4982-25G
APOSOR4982-1GEA 417 CZK
APOSOR4982-1G APOSOR4982-5G APOSOR4982-25G
4-(tert-Butyl)-2-nitroaniline
4-(tert-Butyl)-2-nitroaniline
Vzorec: C₁₀H₁₄N₂O₂
MW: 194,2304 g/mol
Bod tání: 101…105 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00066305
CAS číslo: 6310-19-6
UN: 2811
ADR: 6.1,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR