4-tert-Butylbenzaldehyde 98%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
Synonyms:
4-(tert-Butyl)benzaldehyde
OR4973-100G
OR4973-25G
OR4973-500G
APOSOR4973-100GEA
1240
CZK
APOSOR4973-100G
APOSOR4973-25G
APOSOR4973-500G
4-tert-Butylbenzaldehyde 98%
4-tert-Butylbenzaldehyde
Vzorec:
C₁₁H₁₄O MW: 162,23 g/mol Bod varu: 112…114 °C Hustota: 0,97 g/cm³ (25 °C) Bod vzplanutí: 101 °C Storage Temperature: Ambient |
MDL Number:
MFCD00035742 CAS číslo: 939-97-9 EINECS: 213-367-9 UN: 3082 ADR: 9+EHS,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...