2-(Methylthio)aniline 97%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
Synonyms:
2-(Methylmercapto)aniline
OR4955-25G
OR4955-5G
APOSOR4955-25GEA
1240
CZK
APOSOR4955-25G
APOSOR4955-5G
2-(Methylthio)aniline 97%
2-(Methylthio)aniline
Vzorec:
C₇H₉NS MW: 139,22 g/mol Bod varu: 233…234 °C Hustota: 1,111 g/cm³ (25 °C) Bod vzplanutí: >113 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00007708 CAS číslo: 2987-53-3 EINECS: 221-055-9 UN: 2810 ADR: 6.1,III |
Learn more
![](/m-cz.vwr.com/cs_CZ/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...