2',3',4'-Trihydroxyacetophenone 97%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
1-(2,3,4-Trihydroxyphenyl)ethanone, Gallacetophenone
OR4951-100G
OR4951-25G
APOSOR4951-100GEA
10780
CZK
APOSOR4951-100G
APOSOR4951-25G
2',3',4'-Trihydroxyacetophenone 97%
2',3',4'-Trihydroxyacetophenone
Vzorec:
C₈H₈O₄ MW: 168,15 g/mol Bod tání: 169…172 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002193 CAS číslo: 528-21-2 EINECS: 208-430-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...