3-tert-Butylaniline 97%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
3-(tert-Butyl)aniline, m-tert-Butylaniline
OR4922-25G
OR4922-5G
APOSOR4922-25GEA
6850
CZK
APOSOR4922-25G
APOSOR4922-5G
3-tert-Butylaniline 97%
3-tert-Butylaniline
Vzorec:
C₁₀H₁₅N Bod varu: 126 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00125078 CAS číslo: 5369-19-7 EINECS: 226-361-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...