Tisk…

2-Amino-6-chloropyrimidin-4(3H)-one

Dodavatel: Apollo Scientific

Synonyms: 2-Amino-4-chloro-6-pyrimidinol, 2-Amino-4-chloro-6-hydroxypyrimidine

OR49059-100G OR49059-25G
APOSOR49059-100GEA 0 CZK
APOSOR49059-100G APOSOR49059-25G
2-Amino-6-chloropyrimidin-4(3H)-one
2-Amino-6-chloropyrimidin-4(3H)-one
Vzorec: C₄H₄ClN₃O
MW: 145,5500 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00006093
CAS číslo: 1194-21-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR