Tisk…

4-Chlorothiazole

Dodavatel: Apollo Scientific

OR49049-1G OR49049-5G
APOSOR49049-1GEA 2480 CZK
APOSOR49049-1G APOSOR49049-5G
4-Chlorothiazole
4-Chlorothiazole
Vzorec: C₃H₂ClNS
Storage Temperature: Ambient
MDL Number: MFCD09038604
CAS číslo: 4175-72-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR