4-Aminopiperidine 97%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
Synonyms:
Piperidin-4-amine
OR4858-25G
APOSOR4858-25GEA
2860
CZK
APOSOR4858-25G
4-Aminopiperidine 97%
4-Aminopiperidine
Vzorec:
C₅H₁₂N₂ MW: 100,16 g/mol Bod varu: 65 °C/18mmHg Hustota: 0,945 g/cm³ (25 °C) Bod vzplanutí: 45,6 °C Storage Temperature: Ambient |
MDL Number:
MFCD02179399 CAS číslo: 13035-19-3 UN: 2733 ADR: 3+8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...