Tisk…

Methyl carbazate

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: Methyl hydrazinecarboxylate, Methyl hydrazinocarboxylate

OR4850-100G OR4850-25G
APOSOR4850-100GEA 1210 CZK
APOSOR4850-100G APOSOR4850-25G
Methyl carbazate
Methyl carbazate
Vzorec: C₂H₆N₂O₂
MW: 90,08 g/mol
Bod varu: 108 °C/12mmHg
Bod tání: 70…73 °C
Storage Temperature: Ambient
MDL Number: MFCD00007594
CAS číslo: 6294-89-9
EINECS: 228-560-3
UN: 2811
ADR: 6.1,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR