4-Nitrophthalimide 98%
Dodavatel: Apollo Scientific
Synonyms:
5-Nitroisoindoline-1,3-dione
OR4839-100G
OR4839-25G
OR4839-5G
APOSOR4839-100GEA
8700
CZK
APOSOR4839-100G
APOSOR4839-25G
APOSOR4839-5G
4-Nitrophthalimide 98%
4-Nitrophthalimide
Vzorec:
C₈H₄N₂O₄ MW: 192,13 g/mol Bod tání: 203…205 °C Storage Temperature: Ambient |
MDL Number:
MFCD00005884 CAS číslo: 89-40-7 |
Learn more
![](/m-cz.vwr.com/cs_CZ/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...