Tisk…

2-Chloropyridine-6-carbonitrile 98%

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: 6-Chloropicolinonitrile, 6-Chloropyridine-2-carbonitrile, 2-Chloro-6-cyanopyridine

OR4835-1G OR4835-5G OR4835-10G
APOSOR4835-1GEA 622 CZK
APOSOR4835-1G APOSOR4835-5G APOSOR4835-10G
2-Chloropyridine-6-carbonitrile 98%
2-Chloropyridine-6-carbonitrile
Vzorec: C₆H₃ClN₂
MW: 138,56 g/mol
Bod tání: 85…89 °C
Storage Temperature: Ambient
MDL Number: MFCD00274527
CAS číslo: 33252-29-8
EINECS: 251-429-7
UN: 3439
ADR: 6.1,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR