4-(Prop-2-yn-1-yl)morpholine 98%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
4-(2-Propyn-1-yl)morpholine, 4-Propargylmorpholine
OR48271-1G
OR48271-25G
OR48271-5G
APOSOR48271-1GEA
622
CZK
APOSOR48271-1G
APOSOR48271-25G
APOSOR48271-5G
4-(Prop-2-yn-1-yl)morpholine 98%
4-(Prop-2-yn-1-yl)morpholine
Vzorec:
C₇H₁₁NO MW: 125,17 g/mol Bod varu: 186 °C Storage Temperature: Ambient |
MDL Number:
MFCD00510919 CAS číslo: 5799-76-8 UN: 1993 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...