Tisk…

4-Bromo-2-chlorobenzonitrile

Dodavatel: Apollo Scientific
Nebezpečí

OR48257-100G OR48257-25G OR48257-10G
APOSOR48257-100GEA 3740 CZK
APOSOR48257-100G APOSOR48257-25G APOSOR48257-10G
4-Bromo-2-chlorobenzonitrile
4-Bromo-2-chlorobenzonitrile
Vzorec: C₇H₃BrClN
MW: 216.46
Bod tání: 67…72 °C
Storage Temperature: Ambient
MDL Number: MFCD00040883
CAS číslo: 154607-01-9
UN: 3439
ADR: 6.1 III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR