4-Chloro-3-nitrophenylacetic acid
Dodavatel: Apollo Scientific
|
Nebezpečí
|
OR4823-1G
OR4823-5G
OR4823-25G
APOSOR4823-1GEA
1040
CZK
APOSOR4823-1G
APOSOR4823-5G
APOSOR4823-25G
4-Chloro-3-nitrophenylacetic acid
4-Chloro-3-nitrophenylacetic acid
Vzorec:
C₈H₆ClNO₄ MW: 215,59 g/mol Bod tání: 105…106 °C Storage Temperature: Ambient |
MDL Number:
MFCD00016990 CAS číslo: 37777-68-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...