2-(3-Aminophenyl)acetic acid
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
3-Aminophenylacetic acid
OR4815-10G
OR4815-1G
OR4815-25G
OR4815-5G
APOSOR4815-10GEA
4340
CZK
APOSOR4815-10G
APOSOR4815-1G
APOSOR4815-25G
APOSOR4815-5G
2-(3-Aminophenyl)acetic acid
2-(3-Aminophenyl)acetic acid
Vzorec:
C₈H₉NO₂ MW: 151.1626 g/mol Bod tání: 147…150 °C Storage Temperature: Ambient |
MDL Number:
MFCD00075058 CAS číslo: 14338-36-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...