7-Isoquinolinecarboxylic acid
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
Isoquinoline-7-carboxylic acid
OR48110-250MG
OR48110-100MG
OR48110-1G
APOSOR48110-250MGEA
6450
CZK
APOSOR48110-250MG
APOSOR48110-100MG
APOSOR48110-1G
7-Isoquinolinecarboxylic acid
7-Isoquinolinecarboxylic acid
Vzorec:
C₁₀H₇NO₂ MW: 173,1681 g/mol Bod tání: 295…297 °C Hustota: 0,934 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD10699235 CAS číslo: 221050-96-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...