Tisk…

8-Isoquinolinecarboxylic acid

Dodavatel: Apollo Scientific
Varování

Synonyms: Isoquinoline-8-carboxylic acid

OR48090-1G OR48090-250MG
APOSOR48090-1GEA 2320 CZK
APOSOR48090-1G APOSOR48090-250MG
8-Isoquinolinecarboxylic acid
8-Isoquinolinecarboxylic acid
Vzorec: C₁₀H₇NO₂
MW: 173,1681 g/mol
Bod tání: 263…267 °C
Storage Temperature: Refrigerator
MDL Number: MFCD06738935
CAS číslo: 61563-43-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR