Tisk…

7-Bromo-1H-indole-2-carboxylic acid 95%

Dodavatel: Apollo Scientific

OR480830-1G OR480830-5G
APOSOR480830-1GEA 2940 CZK
APOSOR480830-1G APOSOR480830-5G
7-Bromo-1H-indole-2-carboxylic acid 95%
7-Bromo-1H-indole-2-carboxylic acid
Vzorec: C₉H₆BrNO₂
MW: 240,0500 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00744954
CAS číslo: 16732-71-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR