Tisk…

5-Methylnicotinaldehyde 97%

Dodavatel: Apollo Scientific

Synonyms: 5-Methyl-3-pyridinecarboxaldehyde

OR480820-1G OR480820-5G
APOSOR480820-1GEA 3580 CZK
APOSOR480820-1G APOSOR480820-5G
5-Methylnicotinaldehyde 97%
5-Methylnicotinaldehyde
Vzorec: C7H7NO
MW: 121,1366 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08272078
CAS číslo: 100910-66-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR