Tisk…

4-Methoxycyclohexanone 95%

Dodavatel: Apollo Scientific

OR480813-5G OR480813-25G
APOSOR480813-5GEA 3480 CZK
APOSOR480813-5G APOSOR480813-25G
4-Methoxycyclohexanone 95%
4-Methoxycyclohexanone
Vzorec: C₇H₁₂O₂
Storage Temperature: Ambient
MDL Number: MFCD01110115
CAS číslo: 13482-23-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR