5,6,7,8-Tetrahydroimidazo[1,2-a]pyridine 95%
Dodavatel: Apollo Scientific
|
Varování
|
OR480805-1G
OR480805-250MG
OR480805-500MG
APOSOR480805-1GEA
12670
CZK
APOSOR480805-1G
APOSOR480805-250MG
APOSOR480805-500MG
5,6,7,8-Tetrahydroimidazo[1,2-a]pyridine 95%
5,6,7,8-Tetrahydroimidazo[1,2-a]pyridine
Vzorec:
C₇H₁₀N₂ Storage Temperature: Refrigerator |
MDL Number:
MFCD11849324 CAS číslo: 34167-66-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...