2-Ethynyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 95%
Dodavatel: Apollo Scientific
OR480797-1G
OR480797-250MG
OR480797-5G
APOSOR480797-1GEA
3340
CZK
APOSOR480797-1G
APOSOR480797-250MG
APOSOR480797-5G
2-Ethynyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 95%
2-Ethynyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Vzorec:
C₈H₁₃BO₂ Storage Temperature: Ambient |
MDL Number:
MFCD13182122 CAS číslo: 347389-74-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...