Tisk…

4-Carboxy-3-nitrophenylboronic acid 95%

Dodavatel: Apollo Scientific

OR480791-1G OR480791-5G
APOSOR480791-1GEA 3060 CZK
APOSOR480791-1G APOSOR480791-5G
4-Carboxy-3-nitrophenylboronic acid 95%
4-Carboxy-3-nitrophenylboronic acid
Vzorec: C₇H₆BNO₆
Storage Temperature: Ambient
MDL Number: MFCD10696661
CAS číslo: 80500-28-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR