Tisk…

Ethyl-4-cyano-5-(methylthio)thiophene-2-carboxylate 97%

Dodavatel: Apollo Scientific

Synonyms: 4-Cyano-5-methylsulphanylthiophene-2-carboxylic acid ethyl ester, Ethyl 4-cyano-5-(methylthio)thiophene-2-carboxylate

OR480788-1G OR480788-5G
APOSOR480788-1GEA 2440 CZK
APOSOR480788-1G APOSOR480788-5G
Ethyl-4-cyano-5-(methylthio)thiophene-2-carboxylate 97%
Ethyl-4-cyano-5-(methylthio)thiophene-2-carboxylate
Vzorec: C₉H₉NO₂S₂
MW: 227,31 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01859680
CAS číslo: 116170-84-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR