6-Bromo-2-(methylamino)pyridine 97%
Dodavatel: Apollo Scientific
Synonyms:
6-Bromo-N-methyl-pyridin-2-amine, 6-Bromo-N-methylpyridin-2-amine
OR480763-1G
OR480763-5G
APOSOR480763-1GEA
2610
CZK
APOSOR480763-1G
APOSOR480763-5G
6-Bromo-2-(methylamino)pyridine 97%
6-Bromo-2-(methylamino)pyridine
Vzorec:
C₆H₇BrN₂ MW: 187.0400 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09261221 CAS číslo: 89026-79-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...