Tisk…

N-Boc-3-cyanopyrrolidine 97%

Dodavatel: Apollo Scientific
Varování

Synonyms: N-Boc-3-Cyanopyrrolidine

OR480753-25G OR480753-5G
APOSOR480753-25GEA 15510 CZK
APOSOR480753-25G APOSOR480753-5G
N-Boc-3-cyanopyrrolidine 97%
N-Boc-3-cyanopyrrolidine
Vzorec: C₁₀H₁₆N₂O₂
MW: 196.2462 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03001709
CAS číslo: 476493-40-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR