2-Bromo-5-hydroxybenzonitrile 95%
Dodavatel: Apollo Scientific
|
Varování
|
OR480751-25G
APOSOR480751-25GEA
4470
CZK
APOSOR480751-25G
2-Bromo-5-hydroxybenzonitrile 95%
2-Bromo-5-hydroxybenzonitrile
Vzorec:
C₇H₄BrNO MW: 198,0200 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD08061922 CAS číslo: 189680-06-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...