Tisk…

Boc-Lys(Fmoc)-OH 97%

Dodavatel: Apollo Scientific
Varování

Synonyms: N-epsilon-FMOC-N-alpha-BOC-L-Lysine

OR480747-25G
APOSOR480747-25GEA 2030 CZK
APOSOR480747-25G
Boc-Lys(Fmoc)-OH 97%
Boc-Lys(Fmoc)-OH
Vzorec: C₂₆H₃₂N₂O₆
MW: 468,55 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00062032
CAS číslo: 84624-27-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR