Tisk…

5-Chloro-3-methylbenzo[b]thiophene-2-sulphonamide 97%

Dodavatel: Apollo Scientific

OR480733-25G OR480733-5G
APOSOR480733-25GEA 21730 CZK
APOSOR480733-25G APOSOR480733-5G
5-Chloro-3-methylbenzo[b]thiophene-2-sulphonamide 97%
5-Chloro-3-methylbenzo[b]thiophene-2-sulphonamide
Vzorec: C₉H₈ClNO₂S₂
MW: 261,75 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00210142
CAS číslo: 190659-59-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR