6-Chloropyrimidine-4,5-diamine 95%
Dodavatel: Apollo Scientific
Synonyms:
4,5-Diamino-6-chloropyrimidine, 6-Chloro-4,5-diaminopyrimidine
OR480730-5G
APOSOR480730-5GEA
3020
CZK
APOSOR480730-5G
6-Chloropyrimidine-4,5-diamine 95%
6-Chloropyrimidine-4,5-diamine
Vzorec:
C₄H₅ClN₄ MW: 144,5600 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00023270 CAS číslo: 4316-98-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...