Tisk…

3,6-Dibromothieno[3,2-b]thiophene 98%

Dodavatel: Apollo Scientific

OR480723-25G OR480723-5G
APOSOR480723-25GEA 10150 CZK
APOSOR480723-25G APOSOR480723-5G
3,6-Dibromothieno[3,2-b]thiophene 98%
3,6-Dibromothieno[3,2-b]thiophene
Vzorec: C₆H₂Br₂S₂
MW: 298,01 g/mol
Bod tání: 127 °C
Storage Temperature: Refrigerator
MDL Number: MFCD10000958
CAS číslo: 392662-65-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR