Tisk…

2,6-Dichlorobenzenesulphonamide 95%

Dodavatel: Apollo Scientific

OR480721-25G OR480721-5G
APOSOR480721-25GEA 14450 CZK
APOSOR480721-25G APOSOR480721-5G
2,6-Dichlorobenzenesulphonamide 95%
2,6-Dichlorobenzenesulphonamide
Vzorec: C₆H₅Cl₂NO₂S
Storage Temperature: Ambient
CAS číslo: 10290-98-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR