Tisk…

2,4-Dichloro-5-(chloromethyl)pyrimidine 95%

Dodavatel: Apollo Scientific

OR480717-1G OR480717-5G
APOSOR480717-1GEA 1700 CZK
APOSOR480717-1G APOSOR480717-5G
2,4-Dichloro-5-(chloromethyl)pyrimidine 95%
2,4-Dichloro-5-(chloromethyl)pyrimidine
Vzorec: C₅H₃Cl₃N₂
Storage Temperature: Ambient
MDL Number: MFCD10697639
CAS číslo: 7627-38-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR