Tisk…

Cyclohexyl-4-methoxyphenyl ketone 95%

Dodavatel: Apollo Scientific

OR480695-1G OR480695-5G
APOSOR480695-1GEA 4200 CZK
APOSOR480695-1G APOSOR480695-5G
Cyclohexyl-4-methoxyphenyl ketone 95%
Cyclohexyl-4-methoxyphenyl ketone
Vzorec: C₁₄H₁₈O₂
MW: 218,3 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03841341
CAS číslo: 7469-80-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR