Tisk…

(3-Bromo-4,5-dihydroisoxazol-5-ylmethyl)carbamic acid tert-butyl ester 97%

Dodavatel: Apollo Scientific

OR480678-10G OR480678-2.5G
APOSOR480678-10GEA 16670 CZK
APOSOR480678-10G APOSOR480678-2.5G
(3-Bromo-4,5-dihydroisoxazol-5-ylmethyl)carbamic acid tert-butyl ester 97%
(3-Bromo-4,5-dihydroisoxazol-5-ylmethyl)carbamic acid tert-butyl ester
Vzorec: C₉H₁₅BrN₂O₃
MW: 279,14 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08741480
CAS číslo: 109770-82-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR