Tisk…

Ethyl morpholine-3-carboxylate hydrochloride 90%

Dodavatel: Apollo Scientific

OR480653-1G OR480653-5G
APOSOR480653-1GEA 6750 CZK
APOSOR480653-1G APOSOR480653-5G
Ethyl morpholine-3-carboxylate hydrochloride 90%
Ethyl morpholine-3-carboxylate hydrochloride
Vzorec: C₇H₁₄ClNO₃
MW: 195,65 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12755498
CAS číslo: 84005-98-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR