Tisk…

2-(Carbobenzoxyamino)-1-ethanol 98%

Dodavatel: Apollo Scientific

Synonyms: 2-(Cbz-amino)-1-ethanol, N-Z-Ethanolamine, Benzyl N-(2-Hydroxyethyl)carbamate, Benzyl (2-hydroxyethyl)carbamate, Z-Glycinol, N-(2-Hydroxyethyl)carbamic Acid Benzyl Ester

OR480631-100G
APOSOR480631-100GEA 2480 CZK
APOSOR480631-100G
2-(Carbobenzoxyamino)-1-ethanol 98%
2-(Carbobenzoxyamino)-1-ethanol
Vzorec: C₁₀H₁₃NO₃
MW: 195,22 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00191060
CAS číslo: 77987-49-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR