Tisk…

(4-Iodophenyl)acetic acid 95%

Dodavatel: Apollo Scientific
Varování

Synonyms: 2-(4-Iodophenyl)acetic acid

OR480627-100G OR480627-25G
APOSOR480627-100GEA 7010 CZK
APOSOR480627-100G APOSOR480627-25G
(4-Iodophenyl)acetic acid 95%
(4-Iodophenyl)acetic acid
Vzorec: (C₆H₄I)CH₂COOH
MW: 262,05 g/mol
Bod tání: 136…140 °C
Storage Temperature: Ambient
MDL Number: MFCD00082985
CAS číslo: 1798-06-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR