Tisk…

4-Amino-5,6-dichloropyrimidine 95%

Dodavatel: Apollo Scientific

OR480624-10G
APOSOR480624-10GEA 0 CZK
APOSOR480624-10G
4-Amino-5,6-dichloropyrimidine 95%
4-Amino-5,6-dichloropyrimidine
Vzorec: C₄H₃Cl₂N₃
MW: 163,99 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09837293
CAS číslo: 310400-38-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR