1-Methyl-1,2,3,4-tetrahydroisoquinoline 95%
Dodavatel: Apollo Scientific
OR480616-100G
OR480616-25G
OR480616-50G
APOSOR480616-100GEA
9330
CZK
APOSOR480616-100G
APOSOR480616-25G
APOSOR480616-50G
1-Methyl-1,2,3,4-tetrahydroisoquinoline 95%
1-Methyl-1,2,3,4-tetrahydroisoquinoline
Vzorec:
C₁₀H₁₃N MW: 147,22 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00798989 CAS číslo: 4965-09-7 |
Learn more
![](/m-cz.vwr.com/cs_CZ/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...